ENA336956275 Enamine

N-[(furan-3-yl)methyl]cyclohexanamine

  • MDL number MFCD02177427
  • Cas Number : 462068-58-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85439

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85439
Availability details for
Quantity
  • Availability
  • Add to Cart