ENA336956277 Enamine

(2-chlorophenyl)(3-methoxyphenyl)methanone

  • MDL number MFCD07698791
  • Cas Number : 890098-07-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85441

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85441
Availability details for
Quantity
  • Availability
  • Add to Cart