ENA336956281 Enamine

4-methyl-3-[(phenylcarbamoyl)amino]benzoic acid

  • MDL number MFCD07093101
  • Cas Number : 182957-95-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83115
Availability details for
Quantity
  • Availability
  • Add to Cart