ENA336956282 Enamine

5-(chloromethyl)-3-[(4-fluorophenyl)methyl]-1,2,4-oxadiazole

  • MDL number MFCD09859913
  • Cas Number : 867315-05-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83129

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83129
Availability details for
Quantity
  • Availability
  • Add to Cart