ENA336956290 Enamine

4-fluoro-N-(propan-2-yl)benzene-1-sulfonamide

  • MDL number MFCD01213084
  • Cas Number : 515-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-41816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-41816
Availability details for
Quantity
  • Availability
  • Add to Cart