ENA336956301 Enamine

1-[(3-bromophenyl)methyl]piperidin-2-one

  • MDL number MFCD14661139
  • Cas Number : 1249026-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85487

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85487
Availability details for
Quantity
  • Availability
  • Add to Cart