ENA336956309 Enamine

methyl 2-methyl-1H-imidazole-4-carboxylate

  • MDL number MFCD06203995
  • Cas Number : 97602-72-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136...138
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85495

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-85495
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart