ENA336956312 Enamine

2-(benzyloxy)-1-phenylethan-1-amine

  • MDL number MFCD09729442
  • Cas Number : 127001-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83349

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83349
Availability details for
Quantity
  • Availability
  • Add to Cart