ENA336956317 Enamine

1-cyclopentyl-5-methoxy-2-methyl-1H-indole-3-carboxylic acid

  • MDL number MFCD11890568
  • Cas Number : 1181245-35-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-85504

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85504
Availability details for
Quantity
  • Availability
  • Add to Cart