ENA336956327 Enamine

methyl[(4-methylcyclohexyl)methyl]amine

  • MDL number MFCD18301057
  • Cas Number : 1342108-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83797

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-83797
Availability details for
Quantity
  • Availability
  • Add to Cart