ENA336956331 Enamine

(2-chloroethyl)(methyl)amine hydrochloride

  • MDL number MFCD00050517
  • Cas Number : 4535-90-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 99...101
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83889

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-83889
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart