ENA336956333 Enamine

6-bromo-8-fluoro-3,4-dihydro-2H-1-benzopyran-4-one

  • MDL number MFCD11207542
  • Cas Number : 1092348-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85547

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85547
Availability details for
Quantity
  • Availability
  • Add to Cart