ENA336956334 Enamine

3-(1H-1,2,3,4-tetrazol-5-yl)pentan-3-amine

  • MDL number MFCD12796310
  • Cas Number : 1250299-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85548
Availability details for
Quantity
  • Availability
  • Add to Cart