ENA336956336 Enamine

1-{3-fluoro-4-[methyl(pyridin-2-ylmethyl)amino]phenyl}ethan-1-ol

  • MDL number MFCD12683861
  • Cas Number : 1153369-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 73...75
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85550

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85550
Availability details for
Quantity
  • Availability
  • Add to Cart