ENA336956354 Enamine

2-methanesulfonylcyclopentan-1-one

  • MDL number MFCD15209749
  • Cas Number : 89729-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-85579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85579
Availability details for
Quantity
  • Availability
  • Add to Cart