ENA336956369 Enamine

[(2-bromoethoxy)methyl]cyclopropane

  • MDL number MFCD11136686
  • Cas Number : 56988-04-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83955

Safety Information

signalword Warning
pcodes P210,P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-83955
pictograms GHS07
hstatements H227,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart