ENA336956371 Enamine

2-amino-3-ethoxy-6-fluorobenzoic acid

  • MDL number MFCD17274690
  • Cas Number : 1333681-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-83971

Safety Information

msds https://www.enaminestore.com/catalog/EN300-83971
Availability details for
Quantity
  • Availability
  • Add to Cart