ENA336956378 Enamine

1-{4H,5H,6H,7H-thieno[3,2-c]pyridine-5-carbonyl}piperidine-3-carboxylic acid

  • MDL number MFCD13651002
  • Cas Number : 1096943-94-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-85667

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85667
Availability details for
Quantity
  • Availability
  • Add to Cart