ENA336956382 Enamine

4-chloro-N,N-dimethyl-6-(propan-2-yl)pyrimidin-2-amine

  • MDL number MFCD14607584
  • Cas Number : 71406-67-2
  • Molecular Weight : 199.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84070

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84070
Availability details for
Quantity
  • Availability
  • Add to Cart