ENA336956384 Enamine

(1R)-1-(3-chloro-4-fluorophenyl)ethan-1-ol

  • MDL number MFCD18339573
  • Cas Number : 1344913-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84071

Safety Information

msds https://www.enaminestore.com/catalog/EN300-84071
Availability details for
Quantity
  • Availability
  • Add to Cart