ENA336956385 Enamine

1H,2H,3H-pyrido[2,3-b][1,4]thiazine

  • MDL number MFCD16991714
  • Cas Number : 23977-47-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 127-128
Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85675
Availability details for
Quantity
  • Availability
  • Add to Cart