ENA336956389 Enamine

4-[(benzyloxy)methyl]aniline

  • MDL number MFCD06804489
  • Cas Number : 881741-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85690

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85690
Availability details for
Quantity
  • Availability
  • Add to Cart