ENA336956393 Enamine

[2-(2-chlorophenoxymethyl)phenyl]methanamine

  • MDL number MFCD09906579
  • Cas Number : 951915-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85695

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85695
Availability details for
Quantity
  • Availability
  • Add to Cart