ENA336956395 Enamine

3-(methylsulfanyl)-2-phenylpropanoic acid

  • MDL number MFCD17288065
  • Cas Number : 858208-68-1
  • Molecular Weight : 196.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-85699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85699
Availability details for
Quantity
  • Availability
  • Add to Cart