ENA336956402 Enamine

3-fluoro-1-N-propylbenzene-1,2-diamine

  • MDL number MFCD12149480
  • Cas Number : 1178806-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-45075

Safety Information

msds https://www.enaminestore.com/catalog/EN300-45075
Availability details for
Quantity
  • Availability
  • Add to Cart