ENA336956404 Enamine

pyrimidine-2-sulfonamide

  • MDL number MFCD11847597
  • Cas Number : 142047-90-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85716

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-85716
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart