ENA336956405 Enamine

1-(3-ethoxy-4-nitrophenyl)piperidin-4-ol

  • MDL number MFCD19175034
  • Cas Number : 1089283-25-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109...111
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85717

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85717
Availability details for
Quantity
  • Availability
  • Add to Cart