ENA336956408 Enamine

3-(methanesulfinylmethyl)-1-benzofuran-2-carboxylic acid

  • MDL number MFCD14613368
  • Cas Number : 1250163-72-4
  • Molecular Weight : 238.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-85721

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85721
Availability details for
Quantity
  • Availability
  • Add to Cart