ENA336956414 Enamine

1-[(3-chlorophenyl)methyl]cyclopentane-1-carboxylic acid

  • MDL number MFCD16320492
  • Cas Number : 1226034-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60...62
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85765

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85765
Availability details for
Quantity
  • Availability
  • Add to Cart