ENA336956432 Enamine

2-amino-N,5-dimethylbenzamide

  • MDL number MFCD16783445
  • Cas Number : 34810-84-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-84140

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84140
Availability details for
Quantity
  • Availability
  • Add to Cart