ENA336956434 Enamine

2-hydroxy-2-methyl-3-phenylpropanoic acid

  • MDL number MFCD11052529
  • Cas Number : 56269-86-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90...92
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85800

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85800
Availability details for
Quantity
  • Availability
  • Add to Cart