ENA336956436 Enamine

1-(2-methylbutanoyl)-1,2,3,4-tetrahydroquinoline-6-carboxylic acid

  • MDL number MFCD12463018
  • Cas Number : 1096997-27-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85810
Availability details for
Quantity
  • Availability
  • Add to Cart