ENA336956437 Enamine

ethyl 3-hydroxy-3-(3-hydroxyphenyl)butanoate

  • MDL number MFCD16756852
  • Cas Number : 1249059-47-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85811
Availability details for
Quantity
  • Availability
  • Add to Cart