ENA336956439 Enamine

2,3-dihydro-1H-indene-2-thiol

  • MDL number MFCD12067424
  • Cas Number : 83774-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85832

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85832
Availability details for
Quantity
  • Availability
  • Add to Cart