ENA336956446 Enamine

1-bromo-4-(2-phenylethoxy)benzene

  • MDL number MFCD11123720
  • Cas Number : 30752-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 49...51
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85870

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85870
Availability details for
Quantity
  • Availability
  • Add to Cart