ENA336956448 Enamine

2-cyclohexylcyclopropane-1-carboxylic acid

  • MDL number MFCD16706706
  • Cas Number : 91056-79-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85872
Availability details for
Quantity
  • Availability
  • Add to Cart