ENA336956449 Enamine

2-methyl-3-(thiophen-2-yl)propanoic acid

  • MDL number MFCD07643255
  • Cas Number : 104483-15-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-84151

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84151
Availability details for
Quantity
  • Availability
  • Add to Cart