ENA336956456 Enamine

4-[(6-methylpyridazin-3-yl)oxy]aniline

  • MDL number MFCD13187970
  • Cas Number : 1216239-34-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-85886

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85886
Availability details for
Quantity
  • Availability
  • Add to Cart