ENA336956458 Enamine

7-amino-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD06809628
  • Cas Number : 22246-07-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210...212
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85926

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-85926
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart