ENA336956461 Enamine

3-(1-cyclopropyl-5-fluoro-1H-1,3-benzodiazol-2-yl)-2-methylaniline

  • MDL number MFCD20233655
  • Cas Number : 1354962-68-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85933
Availability details for
Quantity
  • Availability
  • Add to Cart