ENA336956464 Enamine

1-[(4-chlorophenyl)methyl]cyclopentane-1-carboxylic acid

  • MDL number MFCD16320496
  • Cas Number : 1225482-69-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71...73
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85942

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85942
Availability details for
Quantity
  • Availability
  • Add to Cart