ENA336956465 Enamine

3-(thiophen-3-yl)-1,2-oxazol-5-amine

  • MDL number MFCD11156304
  • Cas Number : 1020955-36-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85943

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85943
Availability details for
Quantity
  • Availability
  • Add to Cart