ENA336956474 Enamine

3,4-dimethoxy-N-(2-methoxyethyl)aniline

  • MDL number MFCD11151969
  • Cas Number : 1021000-38-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85955

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85955
Availability details for
Quantity
  • Availability
  • Add to Cart