ENA336956481 Enamine

6-(4-methylpiperazine-1-carbonyl)pyridine-3-carbonitrile

  • MDL number MFCD19180593
  • Cas Number : 1270786-16-7
  • Molecular Weight : 230.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85973
Availability details for
Quantity
  • Availability
  • Add to Cart