ENA336956482 Enamine

2-(2-methylphenyl)morpholine

  • MDL number MFCD09901447
  • Cas Number : 1017395-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-85975
Availability details for
Quantity
  • Availability
  • Add to Cart