ENA336956488 Enamine

1-(3-chlorophenyl)-6-methyl-4-oxo-1,4-dihydropyridazine-3-carboxylic acid

  • MDL number MFCD11544250
  • Cas Number : 68254-09-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 223...225
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-85996

Safety Information

msds https://www.enaminestore.com/catalog/EN300-85996
Availability details for
Quantity
  • Availability
  • Add to Cart