ENA336956501 Enamine

1-bromo-3-ethoxypropane

  • MDL number MFCD07780182
  • Cas Number : 36865-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-86016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86016
Availability details for
Quantity
  • Availability
  • Add to Cart