ENA336956502 Enamine

2-(4-hydroxyphenoxy)propanamide

  • MDL number MFCD11181692
  • Cas Number : 127437-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-86017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86017
Availability details for
Quantity
  • Availability
  • Add to Cart