ENA336956509 Enamine

ethyl 2-amino-4,5-difluorobenzoate

  • MDL number MFCD11226323
  • Cas Number : 864293-36-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-86032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86032
Availability details for
Quantity
  • Availability
  • Add to Cart