ENA336956510 Enamine

2-[2-(trifluoromethyl)phenyl]propanoic acid

  • MDL number MFCD07787628
  • Cas Number : 1080023-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86033
Availability details for
Quantity
  • Availability
  • Add to Cart